Anti-IL-6 Rabbit Recombinant Antibody [clone: RHPM-1 (TOCILIZUMAB)]
Catalog # ABNORAB01172.200UG
Supplier: Abnova
Specifications
- Antigen symbol:IL-6
- Clonality:Recombinant
- Clone:RHPM-1 (TOCILIZUMAB)
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human IL-6.
- Pk:200 µG