Anti-Integrin alpha 4 Rabbit Recombinant Antibody [clone: NATALIZUMAB]
Catalog # ABNORAB01175.200UG
Supplier: Abnova
Specifications
- Antigen symbol:Integrin alpha 4
- Clonality:Recombinant
- Clone:NATALIZUMAB
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human Integrin alpha 4.
- Pk:200 µG