Anti-RSV Mouse Recombinant Antibody [clone: RSHZ19 (FELVIZUMAB)]
Catalog # ABNORAB01178.200UG
Supplier: Abnova
Specifications
- Antigen symbol:RSV
- Clonality:Recombinant
- Clone:RSHZ19 (FELVIZUMAB)
- Host:Mouse
- Immunogen:Recombinant mouse monoclonal antibody raised against RSV.
- Pk:200 µG