Anti-CD3 Mouse Recombinant Antibody [clone: OKT-3 (MUROMONAB)]
Catalog # ABNORAB01138.200UG
Supplier: Abnova
Specifications
- Antigen symbol:CD3
- Clonality:Recombinant
- Clone:OKT-3 (MUROMONAB)
- Host:Mouse
- Reactivity:Human
- Immunogen:Recombinant mouse monoclonal antibody raised against human CD3.
- Pk:200 µG