Anti-CD3 Rabbit Recombinant Antibody [clone: OKT-3 (MUROMONAB)]
Catalog # ABNORAB01139.200UG
Supplier: Abnova
Specifications
- Antigen symbol:CD3
- Clonality:Recombinant
- Clone:OKT-3 (MUROMONAB)
- Host:Rabbit
- Reactivity:Human
- Immunogen:Recombinant rabbit monoclonal antibody raised against human CD3.
- Pk:200 µG