Order Entry
Slovenia
ContactUsLinkComponent
Anti-PRSS21 DNA Rabbit Polyclonal Antibody
Anti-PRSS21 DNA Rabbit Polyclonal Antibody
Catalog # ABNOH00010942-W01P
Supplier:  Abnova
Anti-PRSS21 DNA Rabbit Polyclonal Antibody
Catalog # ABNOH00010942-W01P
Supplier:  Abnova

Specifications

  • Antigen symbol:
    PRSS21 DNA
  • Clonality:
    Polyclonal
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Immunogen:
    Rabbit polyclonal antibody raised against a partial-length human PRSS21 DNA using DNAx™ Immune technology
  • Pk:
    100 µG

Specifications

About this item