Anti-CD11a Rat Monoclonal Antibody [clone: MCD11A]
Catalog # ABNOMAB23631.500UG
Supplier: Abnova
Specifications
- Antigen symbol:CD11a
- Clonality:Monoclonal
- Clone:MCD11A
- Host:Rat
- Reactivity:Mouse
- Immunogen:Rat monoclonal antibody raised against synthetic peptide of Mouse cd11a.
- Pk:500 µG