Anti-CD1a Mouse Monoclonal Antibody [clone: HCD1]
ABNOMAB23578.500UG
: Abnova
- Antigen symbol:CD1a
- Clonality:Monoclonal
- Clone:HCD1
- Host:Mouse
- Reactivity:Human
- Immunogen:Mouse monoclonal antibody raised against synthetic peptide of human CD1a.
- Pk:500 µG