Anti-CD4 Mouse Monoclonal Antibody [clone: HCD4]
ABNOMAB23585.500UG
: Abnova
- Antigen symbol:CD4
- Clonality:Monoclonal
- Clone:HCD4
- Host:Mouse
- Reactivity:Human
- Immunogen:Mouse monoclonal antibody raised against synthetic peptide of human CD4.
- Pk:500 µG