Anti-CD8 Mouse Monoclonal Antibody [clone: HCD8] (Biotin)
ABNOMAB23591.500UG
: Abnova
- Antigen symbol:CD8
- Clonality:Monoclonal
- Clone:HCD8
- Conjugation:Biotin
- Host:Mouse
- Reactivity:Human
- Immunogen:Mouse Biotin monoclonal antibody raised against synthetic peptide of human CD8.
- Pk:500 µG