Anti-CD40 Mouse Monoclonal Antibody [clone: HCD40] (FITC)
Catalog # ABNOMAB23610.500UG
Supplier: Abnova
Specifications
- Antigen symbol:CD40
- Clonality:Monoclonal
- Clone:HCD40
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Mouse
- Reactivity:Human
- Immunogen:Mouse FITC monoclonal antibody raised against synthetic peptide of human CD40.
- Pk:500 µG