Charging base for Rainin AutoRep™ E
Catalog # RAINAR-RB
Supplier: RAININ
Specifications
- Description:Charging base for battery, for AutoRep™ E
- Pk:1
Specifications
About this item
Recharging base for AutoRep™ E battery pack. Accepts complete AutoRep E with battery installed (quicker charge), or battery alone (slower charge).