Glycine ≥98.5% USP
Supplier: Spectrum Chemical
Certificates
About this item
CAS Number:56-40-6
:
Formula:NH₂CH₂COOH
MDL Number:MFCD00008131
:75.07 g/mol
Storage Temperature:Ambient
Specification Test Results
- Assay (NH₂CH₂COOH):98.5 - 101.5%
- Loss On Drying:max. 0.2 %
- Residue On Ignition:max. 0.1 %
- Chloride (Cl):max. 0.007 %
- Sulfates (SO₄):max. 0.0065
Specification Test Results
Frequently Bought Together