Order Entry
Slovenia
ContactUsLinkComponent
Sodium iodide ≥99.0% USP
Irritant
Health
Sodium iodide ≥99.0% USP
Catalog # SPCMSO175-5GM
Supplier:  Spectrum Chemical
CAS Number:  
Irritant
Health
Sodium iodide ≥99.0% USP
Catalog # SPCMSO175-5GM
Supplier:  Spectrum Chemical
CAS Number:  

Specifications

  • Pack type:
    Glass, amber
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Assay (NaI):
    99.0 - 101.5%
  • Alkalinity:
    To Pass Test
  • Water:
    max. 2.0 %
  • Iodate (IO₃):
    To Pass Test
  • Nitrate, Nitrite, and Ammonia:
    To Pass Test

Specification Test Results

About this item

 :
 :149.89 g/mol
Storage Temperature:Ambient
UN:3077
ADR:9,III
 :GHS07
 :GHS08
CAS Number:7681-82-5
Formula:NaI
MDL Number:MFCD00003532