Anti-S100P binding protein Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-6577R-A750
: Bioss
- Antibody type:Primary
- Antigen name:S100P binding protein
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:331-408/408
- Form:Liquid
- Gene ID:64766
- Antigen synonyms:S1P-binding protein|S1PBPR|S1PBP|S1P-binding protein Riken
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:46 kDa
- Storage temperature:Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human S100P binding protein
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
S100PBP was originally cloned from a pancreatic epithelioid carcinoma library and encodes a predicted 408 amino acid protein. RT-PCR detected S100PBP expression in brain, breast, spleen, and lung, but not in pancreas and liver. GFP-tagged S100PBP localised to nuclei of transfected HeLa cells.
Type: Primary
Antigen: S100P binding protein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 331-408/408
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse