Order Entry
Slovenia
ContactUsLinkComponent
Anti-S100P binding protein Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-S100P binding protein Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6577R-A680
Supplier:  Bioss
Anti-S100P binding protein Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-6577R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    S100P binding protein
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    331-408/408
  • Form:
    Liquid
  • Gene ID:
    64766
  • Antigen synonyms:
    S1P-binding protein|S1PBPR|S1PBP|S1P-binding protein Riken
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    46 kDa
  • Storage temperature:
    Store at ‒20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human S100P binding protein
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

S100PBP was originally cloned from a pancreatic epithelioid carcinoma library and encodes a predicted 408 amino acid protein. RT-PCR detected S100PBP expression in brain, breast, spleen, and lung, but not in pancreas and liver. GFP-tagged S100PBP localised to nuclei of transfected HeLa cells.

Type: Primary
Antigen: S100P binding protein
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 331-408/408
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse