Specifications
- Antibody type:Primary
- Antigen name:CPA3/MC-CPA
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Epitope:101-200/417
- Form:Liquid
- Gene ID:1359
- Antigen synonyms:carboxypeptidase A3 mast cell|Mast cell carboxypeptidase A|Cpa3|carboxypeptidase A|MC CPA|Carboxypeptidase A3|mast cell carboxypeptidase A3|MC-CPA|CBPA3_HUMAN
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:36 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human CPA3/MC-CPA
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Carboxypeptidase A (CPA) is a pancreatic exopeptidase which hydrolyses the peptide bond adjacent to the C-terminal end in polypeptide chains. Mast cell carboxypeptidase A (MC-CPA), a part of the peptidase M14 family, is a highly conserved metalloprotease localised to the secretory granules, along with trytases and chymases. MC-CPA is stored as an active enzyme in the granule and is released, along with other inflammatory mediators, upon mast cell degranulation. MC-CPA mirrors pancreatic carboxypeptidase A in cleaving COOH-terminal aromatic and aliphatic amino acid residues. The optimum pH of MC-CPA is between neutral and basic, depending upon the substrate. The MC-CPA gene, CPA3, resides on chromosome 3 and contains 11 exons.
Type: Primary
Antigen: CPA3/MC-CPA
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/417
Host: Rabbit
Isotype: IgG
Reactivity: Rat