Anti-5HT3A Receptor/SR-3A Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-12051R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:5HT3A Receptor/SR-3A
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Epitope:31-130/478
- Form:Liquid
- Gene ID:3359
- Antigen synonyms:5-HT3A|5-hydroxytryptamine receptor 3A|HTR3|5HT3R|5-hydroxytryptamine receptor 3|5-HT-3|5-HT3R|HTR3A|Serotonin-gated ion channel receptor|Serotonin receptor 3A|5-HT3-A
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:53 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human 5HT3A Receptor/SR-3A
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor is a ligand-gated ion channel, which when activated causes fast, depolarizing responses in neurons. It is a cation-specific, but otherwise relatively nonselective, ion channel.
Type: Primary
Antigen: 5HT3A Receptor/SR-3A
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 31-130/478
Host: Rabbit
Isotype: IgG
Reactivity: