Anti-MAOB/Monoamine Oxidase B Rabbit Polyclonal Antibody (Alexa Fluor® 680)
BOSSBS-11297R-A680
: Bioss
- Antibody type:Primary
- Antigen name:MAOB/Monoamine Oxidase B
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Epitope:51-150/520
- Form:Liquid
- Gene ID:4129
- Antigen synonyms:MAOB|Amine oxidase [flavin-containing] B|Monoamine oxidase type B|MAO-B
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:59 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human MAOB/Monoamine Oxidase B
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
catalyses the oxidative deamination of biogenic and xenobiotic amines and has important functions in the metabolism of neuroactive and vasoactive amines in the central nervous system and peripheral tissues. MAOB preferentially degrades benzylamine and phenylethylamine.
Type: Primary
Antigen: MAOB/Monoamine Oxidase B
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 51-150/520
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse