Anti-PDEF/SPDEF Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-1866R-A750
: Bioss
- Antibody type:Primary
- Antigen name:PDEF/SPDEF
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Epitope:251-335/335
- Form:Liquid
- Gene ID:25803
- Antigen synonyms:RP11-375E1__A.3|SPDEF_HUMAN.|PROSTATE EPITHELIUM SPECIC ETS TRANSCRIPTION FACTOR|SAM pointed domain-containing Ets transcription factor|Prostate epithelium-specic Ets transcription factor|PSE|PDEF|Prostate specic Ets|SAM POINTED DOMAIN CONTAINING ETS TRANSCRIPTION FACTOR|Prostate derived Ets factor|Spdef|Prostate-derived Ets factor|Prostate-specic Ets|bA375E1.3
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:38 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human PDEF
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
SPDEF, also known as PDEF is an ETS transcription factor expressed in the prostate epithelial cells. It is thought to act as an androgen-independent transactivator of the prostate-specific antigen (PSA) promoter and also as a transcriptional activator for the SERPINB5 promoter. It is highly expressed in prostate carcinoma cells and is thought to be involved in the regulation of the prostate gland and/or prostate cancer development. It is also expressed in epithelial breast carcinoma.
Type: Primary
Antigen: PDEF/SPDEF
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 251-335/335
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat