Anti-5HT3B receptor Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-4289R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:5HT3B receptor
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Epitope:101-200/441
- Form:Liquid
- Gene ID:9177
- Antigen synonyms:Serotonin receptor 3B|HTR3B|5-HT3B|5-HT3-B|5-hydroxytryptamine receptor 3B
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:50 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human 5HTR3B/5HT3B receptor
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor is a ligand-gated ion channel, which when activated causes fast, depolarising responses. It is a cation-specific, but otherwise relatively nonselective, ion channel.
Type: Primary
Antigen: 5HT3B receptor
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/441
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat