Order Entry
Slovenia
ContactUsLinkComponent
Anti-5HT3B receptor Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Anti-5HT3B receptor Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-4289R-A680
Supplier:  Bioss
Anti-5HT3B receptor Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-4289R-A680
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    5HT3B receptor
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 680
  • Flow cytometry:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat
  • Western blot:
    Yes
  • Epitope:
    101-200/441
  • Form:
    Liquid
  • Gene ID:
    9177
  • Antigen synonyms:
    Serotonin receptor 3B|HTR3B|5-HT3B|5-HT3-B|5-hydroxytryptamine receptor 3B
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    50 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human 5HTR3B/5HT3B receptor
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. This receptor is a ligand-gated ion channel, which when activated causes fast, depolarising responses. It is a cation-specific, but otherwise relatively nonselective, ion channel.

Type: Primary
Antigen: 5HT3B receptor
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/441
Host: Rabbit
Isotype: IgG
Reactivity: Human; Rat