Order Entry
Slovenia
ContactUsLinkComponent
Anti-APNG/MPG Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-APNG/MPG Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-2412R-A750
Supplier:  Bioss
Anti-APNG/MPG Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-2412R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    APNG/MPG
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    101-200/298
  • Form:
    Liquid
  • Gene ID:
    4350
  • Antigen synonyms:
    N methylpurine DNA glycosirase|ADPG|CRA36.1|anpg |N-methylpurine-DNA glycosylase isoform a|AAG|Mid1|MDG|PIG11|3 alkyladenine DNA glycosylase|3MG_HUMAN|Alkyladenine DNA glycosylase|APNG|DNA 3 methyladenine glycosylase|PIG16
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    33 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human APNG/MPG
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

MPG (N-methylpurine DNA glycosylase, or 3-methyladenine (3MeA) DNA glycosylase) repairs the 3MeA lethal lesion blocking DNA replication in Escherichia coli. Analysis of human cDNA libraries found a cDNA that would hybridise to human genomic DNA but not to E. coli or yeast DNA. This MPG gene has been mapped to human chromosome 16 by analysis of a panel of DNAs from mouse/human and hamster/human hybrid cell lines.

Type: Primary
Antigen: APNG/MPG
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/298
Host: Rabbit
Isotype: IgG
Reactivity: Human