Anti-APNG/MPG Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-2412R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:APNG/MPG
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Epitope:101-200/298
- Form:Liquid
- Gene ID:4350
- Antigen synonyms:N methylpurine DNA glycosirase|ADPG|CRA36.1|anpg |N-methylpurine-DNA glycosylase isoform a|AAG|Mid1|MDG|PIG11|3 alkyladenine DNA glycosylase|3MG_HUMAN|Alkyladenine DNA glycosylase|APNG|DNA 3 methyladenine glycosylase|PIG16
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:33 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human APNG/MPG
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
MPG (N-methylpurine DNA glycosylase, or 3-methyladenine (3MeA) DNA glycosylase) repairs the 3MeA lethal lesion blocking DNA replication in Escherichia coli. Analysis of human cDNA libraries found a cDNA that would hybridise to human genomic DNA but not to E. coli or yeast DNA. This MPG gene has been mapped to human chromosome 16 by analysis of a panel of DNAs from mouse/human and hamster/human hybrid cell lines.
Type: Primary
Antigen: APNG/MPG
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 101-200/298
Host: Rabbit
Isotype: IgG
Reactivity: Human