- Antibody type:Primary
- Antigen name:GPR102/TAAR8
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:51-150/342
- Form:Liquid
- Antigen synonyms:Trace amine-associated receptor 8|Trace amine assiciated receptor 8|TaR-8|TAAR8_HUMAN|G-protein coupled receptor 102|TRAR5|GPR102|GPCR GPR102|Trace amine receptor 5|TAR5|Trace amine receptor 8|TAAR8|TA5|G protein coupled receptor 102|TaR-5
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:38 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human G protein coupled receptor 102
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Trace amines are endogenous molecules structurally related to classical biogenic amines that are linked to psychiatric conditions. A family of G-protein coupled receptors referred to as trace-amine-associated receptors (TAAR) are activated by trace amines and are present in very low levels in mammalian tissue. TaRs contain several structural features that are similar to the rhodopsin -adrenergic receptor superfamily, including the positions of the seven transmembrane regions that provide common ligand-binding pockets as well as the short N- and C-terminal domains. TAAR proteins are potential targets for drugs of abuse, such as amphetamine and MDMA, as well as neuropsychiatric disorders including schizophrenia, depression, and attention deficit disorder.
Type: Primary
Antigen: GPR102/TAAR8
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/342
Host: Rabbit
Isotype: IgG
Reactivity: