Anti-c-Fos Rabbit Polyclonal Antibody
Catalog # NOVUNB110-75039
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:c-Fos
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Primates,Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Gene ID:2353
- Antigen synonyms:proto-oncogene c-Fos|AP-1|activator protein 1|G0S7|v-fos FBJ murine osteosarcoma viral oncogene homolog|FBJ murine osteosarcoma viral oncogene homolog|c-fos|FBJ murine osteosarcoma viral (v-fos) oncogene homolog (oncogene FOS)|cellular oncogene c-fos|Cellular oncogene fos|G0/G1 switch regulatory protein 7
- Storage buffer:Whole antisera
- Storage temperature:Store at 4 °C short term. Aliquot and store at ‒20 °C long term. Avoid freeze-thaw cycles.
- Immunogen:A synthetic peptide from aa region 300-380 of human cFos protein conjugated to an immunogenic carrier protein has been used as the antigen. The peptide is homologous with the corresponding sequence derived from cFos protein in many other species including rat, mouse, hamster and cat.
- Purification:Unpurified
- Pk:100 µl
Specifications
About this item
The c-Fos Antibody from Novus Biologicals is a rabbit polyclonal antibody to c-Fos. This antibody reacts with human, mouse, rat, primate. The c-Fos Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:400 to 1:2000
Type: Primary
Antigen: c-Fos
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Primate