Order Entry
Slovenia
ContactUsLinkComponent
Base Materials for Dehydrated Media
Catalog # BLIF4110302
Supplier:  BIOLIFE ITALIANA
CAS Number:  
Base Materials for Dehydrated Media
Catalog # BLIF4110302
Supplier:  BIOLIFE ITALIANA
CAS Number:  
Specifications
  • Pk:
    500 g
  • Description:
    Bios special agar LL
  • Seq:
    23
Specifications
About this item
The ingredients used to prepare culture media are very important, because ultimately the results depend on the quality of these compounds. Raw materials should be stored at 10 to 30 °C unless otherwise stated, in their closed containers in a dry environment.
  • Ingredients are specifically selected to ensure consistent, reproducible results
  • Manufactured under the strictest cGMP standards