Industrial gloves, EDGE® 48-205 series
Supplier: Ansell
Certificates
About this item
An affordable solution for demanding extraction jobs.
- High-performance fibres used in the construction of the palm protect hands from cut injuries
- Back of hand TPR bumpers significantly reduce the risk of crush, pinch, and impact injuries
- Excellent grip performance in dry, wet, and oily environments due to unique material on the palm
- Reinforcement patches between thumb crotch and palm area improve longevity
- Wide cuff with pull strap allows easy donning and doffing
Gloves offer superb cut resistant protection and are suitable for heavy-duty extraction and maintenance work where cut and pinch hazards are present. Bright yellow finish ensures that hands are visible at all times.
Specifications
- Catalog No:
- ANSE48-205/10
- ANSE48-205/11
- ANSE48-205/12
- ANSE48-205/8
- ANSE48-205/9
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex / latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered / powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Finish:
- Vinyl TPR bumper
- Vinyl TPR bumper
- Vinyl TPR bumper
- Vinyl TPR bumper
- Vinyl TPR bumper
- Cuff style:
- Safety cuff
- Safety cuff
- Safety cuff
- Safety cuff
- Safety cuff
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile