Glycine ≥98.5% FCC
SPCMG1041-2.5KG
:
- Pack type:Poly pail
- Pk:2,5 kg
- Seq:4076
- Assay (C₂H₅No₂); Dried Basis):98.5 - 101.5 %
- Lead (Pb):max.5 mg/kg
- Loss On Drying:max. 0.2 %
- Residue On Ignition:max. 0.1 %
- Insoluble Foreign Matter in Amino Acids:To Pass Test
:
:75.07 g/mol
:Ambient
:56-40-6
:NH₂CH₂COOH
:MFCD00008131