Order Entry
Slovenia
ContactUsLinkComponent
Agar No. 2 dehydrated culture media, Mueller Hinton
Agar No. 2 dehydrated culture media, Mueller Hinton
Catalog # HIMEM1084-500G
Supplier:  HIMEDIA
CAS Number:  
Agar No. 2 dehydrated culture media, Mueller Hinton
Catalog # HIMEM1084-500G
Supplier:  HIMEDIA
CAS Number:  
Specifications
  • Pk:
    500 g
  • Description:
    Mueller Hinton agar No. 2
  • Seq:
    01
Specifications
About this item
For testing the susceptibility of common and rapidly growing bacteria using antimicrobial discs by the Bauer-Kirby method, manufactured to contain low levels of thymine, thymidine, calcium, and magnesium.
  • Microbiological culture media
  • 500 g poly bottles