Order Entry
Slovenia
ContactUsLinkComponent
 
 
DTME (dithio-bis-maleimidoethane), Pierce™
  PIER22335
 :  
undefined
 
DTME (dithio-bis-maleimidoethane), Pierce™
  PIER22335
 :  

 

  • Pk:
    50 mg
  • Seq:
    2370

 

 

  • Product Line:
    Pierce™
  • Labeling Method:
    Chemical Labeling
  • Crosslinker Type:
    Homobifunctional
  • Reactive Moiety:
    Maleimide
  • Spacer:
    Cleavable, Long
  • Form:
    Solid
  • Quantity:
    50 mg
  • Type:
    Crosslinker
  • Solubility:
    DMF, DMSO
  • Spacer Arm:
    13.3 Å

 

 

 :
 :312.36 g/mol
 :GHS07
 :Refrigerator
 :71865-37-7
 :C₁₂H₁₂N₂O₄S₂
 :MFCD01863385