Order Entry
Slovenia
ContactUsLinkComponent
[D-Tyr5]-Gonadorelin, Reference Standard
[D-Tyr5]-Gonadorelin, Reference Standard
Catalog # 1296712.
Supplier:  USP
CAS Number:  
[D-Tyr5]-Gonadorelin, Reference Standard
Catalog # 1296712.
Supplier:  USP
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Standards:
    USP reference standard
  • CAS No.:
    53634-21-2
  • Description:
    [D-Tyr5]-Gonadorelin
  • Synonym(s):
    5-Oxo-L-prolyl-L-histidyl-L-tryptophyl-L-seryl-D-tyrosylglycyl-L-leucyl-L-arginyl-L-prolylglycinamide
  • Description:
    [D-Tyr5]-Gonadorelin

Specifications

About this item

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard