Order Entry
Slovenia
ContactUsLinkComponent
Vecuronium Bromide Related Compounds, Reference Standard
Vecuronium Bromide Related Compounds, Reference Standard
Supplier:  USP
Certificates
Vecuronium Bromide Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    13529-31-2
  • Synonym(s):
    3α,17β-diacetyl-oxy-2β, 16β-bispiperidinyl-5α-androstan
  • Formula:
    C₃₃H₅₄N₂O₄
  • Description:
    Vecuronium Bromide Related Compound A

Specifications

Frequently Bought Together