Order Entry
Slovenia
ContactUsLinkComponent
 
Tenofovir Related Compounds, Reference Standard
 :  USP
 
Tenofovir Related Compounds, Reference Standard
 :  USP

 

  • CAS No.:
    1432630-26-6
  • Synonym(s):
    O,O-Bis(isopropoxycarbonyloxymethyl) (S)-[1-(6-amino-9H-purin-9-yl)prop-2-yloxy]methylphosphonate fumarate
  • Formula:
    C₂₃H₃₄N₅O₁₄P
  • Description:
    Tenofovir Disoproxil Related Compound A

 

Frequently Bought Together