Sulfamethoxazole N4-Glucoside, Reference Standard
Catalog # 1631500.
Supplier: USP
CAS Number:
Specifications
- Pack type:Vial
- Pk:25 mg
- Standards:USP reference standard
- CAS No.:119691-75-7
- Description:Sulfamethoxazole N4-Glucoside
- Seq:05
- Synonym(s):4-(β-D-glucopyranosylamino)-N-(5-methyl-3-isoxazolyl)-benzenesulfonamide
- Formula:C₁₆H₂₁N₃O₈S
- Description:Sulfamethoxazole N4-Glucoside
Specifications
About this item
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard