Order Entry
Slovenia
ContactUsLinkComponent
Pantoprazole Sodium, Reference Standard
Pantoprazole Sodium, Reference Standard
Catalog # 1494895.
Supplier:  USP
CAS Number:  
Pantoprazole Sodium, Reference Standard
Catalog # 1494895.
Supplier:  USP
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    250 mg
  • Standards:
    USP reference standard
  • CAS No.:
    164579-32-2
  • Description:
    Pantoprazole Sodium
  • Seq:
    01
  • Formula:
    C₃₂H₃₄F₄N₆Na₂O₁₁S₂
  • Description:
    Pantoprazole Sodium

Specifications

About this item

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard