Order Entry
Slovenia
ContactUsLinkComponent
Nicotine Bitartrate Dihydrate, Reference Standard
Nicotine Bitartrate Dihydrate, Reference Standard
Catalog # 1463304.
Supplier:  USP
CAS Number:  
Nicotine Bitartrate Dihydrate, Reference Standard
Catalog # 1463304.
Supplier:  USP
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    400 mg
  • Standards:
    USP reference standard
  • CAS No.:
    6019-06-3
  • Description:
    Nicotine Bitartrate Dihydrate
  • UN No.:
    1659
  • Formula:
    C₁₈H₃₀N₂O₁₄
  • Description:
    Nicotine Bitartrate Dihydrate

Specifications

About this item

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard