Order Entry
Slovenia
ContactUsLinkComponent
Mirtazapine Related Compounds, Reference Standard
Mirtazapine Related Compounds, Reference Standard
Supplier:  USP
Certificates
Mirtazapine Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    191546-96-0
  • Synonym(s):
    2-Methyl-3,4,10,14b-tetrahydrobenzo[c]pyrazino[1,2-a]pyrido[3,2-f]azepin-1(2H)-one
  • Formula:
    C₁₇H₁₇N₃O
  • Description:
    Mirtazapine Related Compound C

Specifications

Frequently Bought Together