Order Entry
Slovenia
ContactUsLinkComponent
 
Levofloxacin Related Compounds, Reference Standard
 :  USP
 
Levofloxacin Related Compounds, Reference Standard
 :  USP

 

  • CAS No.:
    1346603-62-0
  • Synonym(s):
    (S)-9-Fluoro-2,3-dihydro-3-methyl-10-[2-(methylamino)ethylamino]-7-oxo-7H-pyrido[1,2,3-de][1,4]benzoxazine-6-carboxylic acid hydrochloride
  • Formula:
    C₁₆H₁₉ClFN₃O₄
  • Description:
    Levofloxacin Related Compound E

 

Frequently Bought Together