Order Entry
Slovenia
ContactUsLinkComponent
Granisetron Related Compounds, Reference Standard
Granisetron Related Compounds, Reference Standard
Supplier:  USP
Certificates
Granisetron Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    107007-95-4
  • Synonym(s):
    N-[(1R,3r,5S)-9-methyl-9-azabicyclo[3.3.1]non-3-yl]-1H-indazole-3-carboxamide
  • Formula:
    C₁₇H₂₂N₄O
  • Description:
    Granisetron Related Compound B

Specifications

Frequently Bought Together