Order Entry
Slovenia
ContactUsLinkComponent
Fluticasone Propionate Related Compounds, Reference Standard
Fluticasone Propionate Related Compounds, Reference Standard
Supplier:  USP
Certificates
Fluticasone Propionate Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    28416-82-2
  • UN No.:
    3077
  • Synonym(s):
    6α, 9α-difluoro-11β,17α-dihydroxy-16α-methyl-3-oxoandrosta-1,4-diene-17β-carboxylic acid
  • Formula:
    C₂₁H₂₆F₂O₅
  • Description:
    Fluticasone Propionate Related Compound J

Specifications

Frequently Bought Together