Order Entry
Slovenia
ContactUsLinkComponent
Cyclophosphamide Related Compounds, Reference Standard
Cyclophosphamide Related Compounds, Reference Standard
Supplier:  USP
Certificates
Cyclophosphamide Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    821-48-7
  • Synonym(s):
    bis(2-chloroethyl)amine hydrochloride
  • Description:
    Cyclophosphamide Related Compound A
  • Formula:
    C₃H₁₀NO₄P

Specifications

Frequently Bought Together