Order Entry
Slovenia
ContactUsLinkComponent
Clomipramine Hydrochloride, Reference Standard
Clomipramine Hydrochloride, Reference Standard
Catalog # 1140247.
Supplier:  USP
CAS Number:  
Clomipramine Hydrochloride, Reference Standard
Catalog # 1140247.
Supplier:  USP
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    200 mg
  • Standards:
    USP reference standard
  • CAS No.:
    17321-77-6
  • Description:
    Clomipramine Hydrochloride
  • Seq:
    01
  • UN No.:
    3077
  • Formula:
    C₁₉H₂₄Cl₂N₂
  • Description:
    Clomipramine Hydrochloride

Specifications

About this item

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.

Certifications: US Pharmacopea Reference Standard