Order Entry
Slovenia
ContactUsLinkComponent
Ciclopirox Related Compounds, Reference Standard
Ciclopirox Related Compounds, Reference Standard
Supplier:  USP
Certificates
Ciclopirox Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    14818-35-0
  • Synonym(s):
    6-Cyclohexyl-4-methyl-2-pyrone
  • Formula:
    C₁₂H₁₆O₂
  • Description:
    Ciclopirox Related Compound B

Specifications

Frequently Bought Together