Cetirizine Hydrochloride, Reference Standard
1102929.
: USP
:
- Pack type:Vial
- Pk:250 mg
- Standards:USP reference standard
- CAS No.:83881-52-1
- Description:Cetirizine Hydrochloride
- Seq:01
- Formula:C₂₁H₂₇Cl₃N₂O₃
- Description:Cetirizine Hydrochloride
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard