Order Entry
Slovenia
ContactUsLinkComponent
Atomoxetine Related Compounds, Reference Standard
Atomoxetine Related Compounds, Reference Standard
Supplier:  USP
Certificates
Atomoxetine Related Compounds, Reference Standard
Supplier:  USP

Specifications

  • CAS No.:
    42142-52-9
  • Synonym(s):
    3-(Methylamino)-1-phenylpropan-1-ol
  • Description:
    Atomoxetine Related Compound A

Specifications

Frequently Bought Together