Clopidogrel-Beta-D-glucuronide, Pharmaceutical Analytical Impurities
1A01360.
: USP
:
- Pack type:Vial
- Pk:10 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:1314116-53-4
- Description:Clopidogrel-Beta-D-glucuronide
- Synonym(s):(2S,3S,4S,5R,6S)-6-((S)-2-(2-chlorophenyl)-2-(6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl)acetoxy)-3,4,5-trihydroxytetrahydro-2H-pyran-2-carboxylic acid
- Description:Clopidogrel-Beta-D-glucuronide
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.