Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

Clopidogrel, Pharmaceutical Analytical Impurities

Supplier:   USP
Clopidogrel, Pharmaceutical Analytical Impurities
Supplier:  USP
Certificates

2 Options Available Below

Product Details & Documents

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 1A01380.
  • 1A01870.
  • Description:
  • Clopidogrel Quaternary Salt
  • Clopidogrel R-Enantiomer
  • Synonym(s):
  • 5-(1-(2-chlorophenyl)-2-methoxy-2-oxoethyl)-6,7-dihydrothieno[3,2-c]pyridin-5-ium, bromide
  • (R)-2-(2-chlorophenyl)-2-(6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl)acetamide
  • CAS No.:
  • 2418591-12-3
  • 444728-15-8

Specifications

Product Family Options

Product Information

  • Standards
    Pack type
    Pk
    Availability
    Price
  • 1A01380.
    Clopidogrel Quaternary Salt
    Clopidogrel Quaternary Salt
    Pharmaceutical Analytical Impurities
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 230,00 €
    1 230,00 €
    1 * 25 mg
       1 230,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1A01380.
    Description
    Clopidogrel Quaternary Salt
    Synonym(s)
    5-(1-(2-chlorophenyl)-2-methoxy-2-oxoethyl)-6,7-dihydrothieno[3,2-c]pyridin-5-ium, bromide
    CAS No.
    2418591-12-3
  • 1A01870.
    Clopidogrel R-Enantiomer
    Clopidogrel R-Enantiomer
    Pharmaceutical Analytical Impurities
    Vial
    25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 1 100,00 €
    1 100,00 €
    1 * 25 mg
       1 100,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    1A01870.
    Description
    Clopidogrel R-Enantiomer
    Synonym(s)
    (R)-2-(2-chlorophenyl)-2-(6,7-dihydrothieno[3,2-c]pyridin-5(4H)-yl)acetamide
    CAS No.
    444728-15-8