Clopidogrel N-Oxide, Pharmaceutical Analytical Impurities
Catalog # 1A01890.
Supplier: USP
CAS Number:
Specifications
- Pack type:Vial
- Pk:25 mg
- Standards:Pharmaceutical Analytical Impurities
- CAS No.:1319197-71-1
- Description:Clopidogrel N-Oxide
- Synonym(s):5-((S)-1-(2-chlorophenyl)-2-methoxy-2-oxoethyl)-4,5,6,7-tetrahydrothieno[3,2-c]pyridine 5-oxide
- Description:Clopidogrel N-Oxide
Specifications
About this item
Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.