Order Entry
Slovenia
ContactUsLinkComponent
Tetrahydrothienopyridine Hydrochloride, Pharmaceutical Analytical Impurities
Tetrahydrothienopyridine Hydrochloride, Pharmaceutical Analytical Impurities
Catalog # 1A01850.
Supplier:  USP
CAS Number:  
Tetrahydrothienopyridine Hydrochloride, Pharmaceutical Analytical Impurities
Catalog # 1A01850.
Supplier:  USP
CAS Number:  

Specifications

  • Pack type:
    Vial
  • Pk:
    25 mg
  • Standards:
    Pharmaceutical Analytical Impurities
  • CAS No.:
    28783-41-7
  • Description:
    Tetrahydrothienopyridine Hydrochloride
  • Synonym(s):
    4,5,6,7-Tetrahydrothieno[3,2-c]pyridine, hydrochloride
  • Description:
    Tetrahydrothienopyridine Hydrochloride

Specifications

About this item

Pharmaceutical Analytical Impurities are released through a USP quality process designed to ensure identity and quality appropriate for analytical applications. These impurities are supported with a product information sheet for each batch.